4-Methyl-3-(trifluoromethyl)phenol 98%

Supplier: Apollo Scientific
Danger

PC302756-1G PC302756-5G PC302756-10G
APOSPC302756-1GEA 203 EUR
APOSPC302756-1G APOSPC302756-5G APOSPC302756-10G
4-Methyl-3-(trifluoromethyl)phenol 98%
4-Methyl-3-(trifluoromethyl)phenol
Formula: C₈H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD09832294
CAS Number: 1017778-16-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR