5-Chloro-2-(trifluoromethoxy)benzylamine 98%

Supplier: Apollo Scientific

PC302759-1G PC302759-5G PC302759-250MG
APOSPC302759-1GEA 400 EUR
APOSPC302759-1G APOSPC302759-5G APOSPC302759-250MG
5-Chloro-2-(trifluoromethoxy)benzylamine 98%
5-Chloro-2-(trifluoromethoxy)benzylamine
Formula: C₈H₇ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD11519365
CAS Number: 874821-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR