3-Bromo-5-(trifluoromethoxy)benzoyl chloride 97%

Supplier: Apollo Scientific

PC302826-5G PC302826-1G
APOSPC302826-5GEA 433 EUR
APOSPC302826-5G APOSPC302826-1G
3-Bromo-5-(trifluoromethoxy)benzoyl chloride 97%
3-Bromo-5-(trifluoromethoxy)benzoyl chloride
Formula: C₈H₃BrClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11519379
CAS Number: 1092461-36-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR