3-Methoxy-4-(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific

PC302850-5G PC302850-1G PC302850-25G
APOSPC302850-5GEA 84.1 EUR
APOSPC302850-5G APOSPC302850-1G APOSPC302850-25G
3-Methoxy-4-(trifluoromethyl)benzoic acid 97%
3-Methoxy-4-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD13185682
CAS Number: 276861-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR