2-Chloro-3-(trifluoromethyl)phenylacetonitrile 98%

Supplier: Apollo Scientific
Danger

PC302908-10G PC302908-5G PC302908-1G
APOSPC302908-10GEA 288 EUR
APOSPC302908-10G APOSPC302908-5G APOSPC302908-1G
2-Chloro-3-(trifluoromethyl)phenylacetonitrile 98%
2-Chloro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD04115991
CAS Number: 22902-81-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR