2-Chloro-4-(trifluoromethyl)phenylacetonitrile 97%

Supplier: Apollo Scientific

PC303144-1G PC303144-5G
APOSPC303144-1GEA 264 EUR
APOSPC303144-1G APOSPC303144-5G
2-Chloro-4-(trifluoromethyl)phenylacetonitrile 97%
2-Chloro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD14155640
CAS Number: 474024-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR