(2-Chloro-4-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

PC303152-1G PC303152-5G PC303152-25G
APOSPC303152-1GEA 66.7 EUR
APOSPC303152-1G APOSPC303152-5G APOSPC303152-25G
(2-Chloro-4-(trifluoromethyl)phenyl)acetic acid 98%
(2-Chloro-4-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13194288
CAS Number: 601513-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR