3-Chloro-2,6-difluorobenzenesulphonyl chloride

Supplier: Apollo Scientific
Danger

PC303185-1G
APOSPC303185-1GEA 843 EUR
APOSPC303185-1G
3-Chloro-2,6-difluorobenzenesulphonyl chloride
3-Chloro-2,6-difluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₂F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD12922595
CAS Number: 1208077-31-9
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR