2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide 97%

Supplier: Apollo Scientific

PC303193-1G PC303193-250MG
APOSPC303193-1GEA 731 EUR
APOSPC303193-1G APOSPC303193-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzyl bromide
Formula: C₉H₇BrF₄
Storage Temperature: Ambient
MDL Number: MFCD28103367
CAS Number: 1824048-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR