4-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride

Supplier: Apollo Scientific
Danger

PC303267-1G
APOSPC303267-1GEA 1740 EUR
APOSPC303267-1G
4-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride
4-Chloro-2-fluoro-3-methoxybenzenesulphonyl chloride
Formula: C₇H₅Cl₂FO₃S
MW: 259,082 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054240
CAS Number: 1706435-13-3
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR