5-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde 97%

Supplier: Apollo Scientific

PC303540-10G PC303540-1G PC303540-5G
APOSPC303540-10GEA 993 EUR
APOSPC303540-10G APOSPC303540-1G APOSPC303540-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD18837610
CAS Number: 1291487-26-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR