3-Amino-6-(trifluoromethyl)pyridazine

Supplier: Apollo Scientific
Warning

Synonyms: 6-(Trifluoromethyl)pyridazin-3-amine

PC305008-500MG
APOSPC305008-500MGEA 1180 EUR
APOSPC305008-500MG
3-Amino-6-(trifluoromethyl)pyridazine
3-Amino-6-(trifluoromethyl)pyridazine
Formula: C₅H₄F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD13189559
CAS Number: 935777-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR