(5-Fluoropyridin-2-yl)methanol

Supplier: Apollo Scientific

PC305009-10G PC305009-1G PC305009-5G
APOSPC305009-10GEA 400 EUR
APOSPC305009-10G APOSPC305009-1G APOSPC305009-5G
(5-Fluoropyridin-2-yl)methanol
(5-Fluoropyridin-2-yl)methanol
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD09027049
CAS Number: 802325-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR