α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

PC3060-100G PC3060-500G PC3060-1KG
APOSPC3060-100GEA 29 EUR
APOSPC3060-100G APOSPC3060-500G APOSPC3060-1KG
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine
Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 85 °C/15mmHg
Melting Pt: 3
Density: 1.473 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR