α,α,α,α',α',α'-Hexafluoro-m-xylene 99%

Supplier: Apollo Scientific
Warning

Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene

PC3070-1KG PC3070-250G
APOSPC3070-1KGEA 92.8 EUR
APOSPC3070-1KG APOSPC3070-250G
α,α,α,α',α',α'-Hexafluoro-m-xylene 99%
α,α,α,α',α',α'-Hexafluoro-m-xylene
Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –35 °C
Density: 1.378 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00000392
CAS Number: 402-31-3
EINECS: 206-939-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR