3,5-Bis(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

PC3090-100G PC3090-10G PC3090-5G PC3090-25G
APOSPC3090-100GEA 87 EUR
APOSPC3090-100G APOSPC3090-10G APOSPC3090-5G APOSPC3090-25G
3,5-Bis(trifluoromethyl)benzoic acid 97%
3,5-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
Melting Pt: 140…144 °C
Density: 1,42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000388
CAS Number: 725-89-3
EINECS: 211-970-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR