3,5-Bis(trifluoromethyl)benzonitrile 97%

Supplier: Apollo Scientific
Warning

PC3100-100G PC3100-25G PC3100-500G
APOSPC3100-100GEA 77 EUR
APOSPC3100-100G APOSPC3100-25G APOSPC3100-500G
3,5-Bis(trifluoromethyl)benzonitrile 97%
3,5-Bis(trifluoromethyl)benzonitrile
Formula: C₉H₃F₆N
Boiling Pt: 154…156 °C
Melting Pt: 16…21 °C
Density: 1,421 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000379
CAS Number: 27126-93-8
EINECS: 248-240-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR