2,4-Bis(trifluoromethyl)-DL-phenylalanine 98%

Supplier: Apollo Scientific

PC3114K-1G PC3114K-5G
APOSPC3114K-1GEA 116 EUR
APOSPC3114K-1G APOSPC3114K-5G
2,4-Bis(trifluoromethyl)-DL-phenylalanine 98%
2,4-Bis(trifluoromethyl)-DL-phenylalanine
Formula: C₁₁H₉F₆NO₂
Storage Temperature: Ambient
MDL Number: MFCD00061306
CAS Number: 237076-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR