1H,1H,7H-Dodecafluoroheptyl acrylate 97%

Supplier: Apollo Scientific
Warning

PC3132-100G PC3132-25G
APOSPC3132-100GEA 0 EUR
APOSPC3132-100G APOSPC3132-25G
1H,1H,7H-Dodecafluoroheptyl acrylate 97%
1H,1H,7H-Dodecafluoroheptyl acrylate
Formula: C₁₀H₆F₁₂O₂
Boiling Pt: 197 °C
Density: 1,581 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080746
CAS Number: 2993-85-3
EINECS: 221-064-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR