3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone

Supplier: Apollo Scientific

Synonyms: 3-(4-Methoxyphenyl)-1,1,1-trifluoroacetone

PC32341-1G PC32341-5G
APOSPC32341-1GEA 364 EUR
APOSPC32341-1G APOSPC32341-5G
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone
3-(4-Methoxyphenyl)-1,1,1-trifluoro-2-propanone
Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764734
CAS Number: 22102-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR