Dimethyl (4-fluorobenzyl)malonate

Supplier: Apollo Scientific

PC32416-25G PC32416-5G PC32416-1G
APOSPC32416-25GEA 864 EUR
APOSPC32416-25G APOSPC32416-5G APOSPC32416-1G
Dimethyl (4-fluorobenzyl)malonate
Dimethyl (4-fluorobenzyl)malonate
Formula: C₁₂H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD01934558
CAS Number: 252938-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR