1-Bromo-3-(difluoromethoxy)benzene 98%

Supplier: Apollo Scientific
Warning

PC3429-1G PC3429-5G PC3429-10G
APOSPC3429-1GEA 21.9 EUR
APOSPC3429-1G APOSPC3429-5G APOSPC3429-10G
1-Bromo-3-(difluoromethoxy)benzene 98%
1-Bromo-3-(difluoromethoxy)benzene
Formula: C₇H₅BrF₂O
Boiling Pt: 196…197 °C
Density: 1,585 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236233
CAS Number: 262587-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR