Ethyl-3,5-bis(trifluoromethyl)benzoate

Supplier: Apollo Scientific

PC3553-1G PC3553-5G
APOSPC3553-1GEA 75.9 EUR
APOSPC3553-1G APOSPC3553-5G
Ethyl-3,5-bis(trifluoromethyl)benzoate
Ethyl 3,5-bis(trifluoromethyl)benzoate
Formula: C₁₁H₈F₆O₂
MW: 286.17 g/mol
Boiling Pt: 84 °C/10mmHg
Storage Temperature: Ambient
MDL Number: MFCD01320684
CAS Number: 96617-71-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR