4-Fluorobenzyl alcohol 95+%

Supplier: Apollo Scientific
Warning

Synonyms: (4-Fluorophenyl)methanol

PC3660-100G
APOSPC3660-100GEA 55.1 EUR
APOSPC3660-100G
4-Fluorobenzyl alcohol 95+%
4-Fluorobenzyl alcohol
Formula: C₇H₇FO
Boiling Pt: 209 °C/730mmHg
Melting Pt: 23 °C
Density: 1,156 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004651
CAS Number: 459-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR