4-Fluorobenzyl isothiocyanate

Supplier: Apollo Scientific
Warning

Synonyms: 4-Fluorobenzylisothiocyanate, 1-Fluoro-4-(isothiocyanatomethyl)benzene

PC3721N-5G
APOSPC3721N-5GEA 85.8 EUR
APOSPC3721N-5G
4-Fluorobenzyl isothiocyanate
4-Fluorobenzyl isothiocyanate
Formula: C₈H₆FNS
Boiling Pt: 124…125 °C/6mmHg
Density: 1,22 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041115
CAS Number: 2740-88-7
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR