Ethyl-5-(trifluoromethyl)picolinate

Supplier: Apollo Scientific
Danger

Synonyms: Ethyl 5-(trifluoromethyl)-2-pyridinecarboxylate

PC3723-250MG
APOSPC3723-250MGEA 69.8 EUR
APOSPC3723-250MG
Ethyl-5-(trifluoromethyl)picolinate
Ethyl 5-(trifluoromethyl)picolinate
Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100197
CAS Number: 128072-94-6
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR