Tetrafluoroboric acid min 48% in aqueous solution

Supplier: Apollo Scientific
Danger

Synonyms: Fluoboric acid

PC3728-2.5KG PC3728-500G
APOSPC3728-2.5KGEA 0 EUR
APOSPC3728-2.5KG APOSPC3728-500G
Tetrafluoroboric acid min 48% in aqueous solution
Tetrafluoroboric acid
Formula: HBF₄
Boiling Pt: 130 °C
Density: 1,41 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00011345
CAS Number: 16872-11-0
UN: 1775
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR