(3-Fluoro-4-methoxyphenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-4-methoxyphenylacetic acid

PC3802-100G PC3802-25G PC3802-5G
APOSPC3802-100GEA 156 EUR
APOSPC3802-100G APOSPC3802-25G APOSPC3802-5G
(3-Fluoro-4-methoxyphenyl)acetic acid 98%
(3-Fluoro-4-methoxyphenyl)acetic acid
Formula: C₉H₉FO₃
MW: 184.17 g/mol
Melting Pt: 110…116 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060353
CAS Number: 452-14-2
EINECS: 610-231-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR