1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate 97%

Supplier: Apollo Scientific
Danger

Synonyms: Trifluoromethanesulphonic Acid 1,1,1,3,3,3-Hexafluoroisopropyl Ester, 1,1,1,3,3,3-Hexafluoroisopropyl Triflate

PC3808-1G PC3808-5G
APOSPC3808-1GEA 53.8 EUR
APOSPC3808-1G APOSPC3808-5G
1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate 97%
1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate
Formula: C₄HF₉O₃S
MW: 300.09 g/mol
Boiling Pt: 77…78 °C
Density: 1.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093340
CAS Number: 156241-41-7
UN: 2927
ADR: 6.1+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR