2-Fluoro-5-methylbenzenesulphonyl chloride

Supplier: Apollo Scientific
Danger

PC3871-1G PC3871-25G PC3871-5G
APOSPC3871-1GEA 21.9 EUR
APOSPC3871-1G APOSPC3871-25G APOSPC3871-5G
2-Fluoro-5-methylbenzenesulphonyl chloride
2-Fluoro-5-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
Melting Pt: 30…34 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04039785
CAS Number: 870704-14-6
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR