5-Bromo-2,4-difluorobenzenesulphonyl chloride 97%

Supplier: Apollo Scientific
Danger

PC3924-1G PC3924-25G PC3924-5G PC3924-100G
APOSPC3924-1GEA 21.9 EUR
APOSPC3924-1G APOSPC3924-25G APOSPC3924-5G APOSPC3924-100G
5-Bromo-2,4-difluorobenzenesulphonyl chloride 97%
5-Bromo-2,4-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD01940424
CAS Number: 287172-61-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR