2-Fluoro-4-nitrophenol 98%

Supplier: Apollo Scientific
Warning

PC3969-100G PC3969-25G PC3969-500G
APOSPC3969-100GEA 60.9 EUR
APOSPC3969-100G APOSPC3969-25G APOSPC3969-500G
2-Fluoro-4-nitrophenol 98%
2-Fluoro-4-nitrophenol
Formula: C₆H₄FNO₃
Melting Pt: 120…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00051970
CAS Number: 403-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR