1-Bromo-3-(3,3-difluorocyclobutyl)benzene

Supplier: Apollo Scientific
Warning

PC400053-250MG
APOSPC400053-250MGEA 386 EUR
APOSPC400053-250MG
1-Bromo-3-(3,3-difluorocyclobutyl)benzene
1-Bromo-3-(3,3-difluorocyclobutyl)benzene
Formula: C₁₀H₉BrF₂
Storage Temperature: Ambient
MDL Number: MFCD31621118
CAS Number: 1897738-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR