4-(Difluoromethoxy)pyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Difluoromethoxy)picolinic acid

PC400110-100MG PC400110-250MG
APOSPC400110-100MGEA 422 EUR
APOSPC400110-100MG APOSPC400110-250MG
4-(Difluoromethoxy)pyridine-2-carboxylic acid
4-(Difluoromethoxy)pyridine-2-carboxylic acid
Formula: C₇H₅F₂NO₃
MW: 189.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27995942
CAS Number: 1707365-38-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR