3-(Trifluoromethyl)-1H-pyrazole

Supplier: Apollo Scientific

PC400547-1G
APOSPC400547-1GEA 939 EUR
APOSPC400547-1G
3-(Trifluoromethyl)-1H-pyrazole
3-(Trifluoromethyl)-1H-pyrazole
Formula: C₄H₃F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD00115018
CAS Number: 20154-03-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR