2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine

Supplier: Apollo Scientific

PC400570-1G
APOSPC400570-1GEA 1150 EUR
APOSPC400570-1G
2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine
2-(3-Bromo-2-chloro-6-fluorophenyl)-3H-imidazo[4,5-c]pyridine
Formula: C₁₂H₆BrClFN₃
Storage Temperature: Ambient
MDL Number: MFCD22690833
CAS Number: 1439824-02-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR