(E)-4-(2-Trifluoromethyl-phenyl)-but-3-en-2-one

Supplier: Apollo Scientific
Warning

Synonyms: 2-(Trifluoromethyl)benzylideneacetone

PC401022-1G PC401022-2.5G PC401022-5G PC401022-10G
APOSPC401022-1GEA 209 EUR
APOSPC401022-1G APOSPC401022-2.5G APOSPC401022-5G APOSPC401022-10G
(E)-4-(2-Trifluoromethyl-phenyl)-but-3-en-2-one
(E)-4-(2-Trifluoromethyl-phenyl)-but-3-en-2-one
Formula: C₁₁H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD12827896
CAS Number: 76293-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR