4-(2-Chloro-5-fluorophenyl)piperidine hydrochloride 95+%

Supplier: Apollo Scientific
Warning

PC402112-250MG
APOSPC402112-250MGEA 168 EUR
APOSPC402112-250MG
4-(2-Chloro-5-fluorophenyl)piperidine hydrochloride 95+%
4-(2-Chloro-5-fluorophenyl)piperidine hydrochloride
Formula: C₁₁H₁₃ClFN·ClH
Storage Temperature: Ambient
MDL Number: MFCD31629947
CAS Number: 1821329-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR