tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate 97%

Supplier: Apollo Scientific
Warning

PC402120-1G
APOSPC402120-1GEA 232 EUR
APOSPC402120-1G
tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate 97%
tert-Butyl 4-((2-(4-fluoro-3-(trifluoromethyl)phenyl)-2-oxoethyl)carbamoyl)piperidine-1-carboxylate
Formula: C₂₀H₂₄F₄N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD15071415
CAS Number: 1082949-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR