4-Bromo-3-chloro-7-fluoroquinoline

Supplier: Apollo Scientific

PC404603-1G
APOSPC404603-1GEA 525 EUR
APOSPC404603-1G
4-Bromo-3-chloro-7-fluoroquinoline
4-Bromo-3-chloro-7-fluoroquinoline
Formula: C₉H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD13619700
CAS Number: 1208814-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR