(+/-)-3-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid

Supplier: Apollo Scientific

PC405604-100MG
APOSPC405604-100MGEA 1300 EUR
APOSPC405604-100MG
(+/-)-3-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
(+/-)-3-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
Formula: C₁₁H₉F₃O₂
MW: 230.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683236
CAS Number: 1404192-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR