3-(Pentafluorothio)thiophenol

Supplier: Apollo Scientific
Warning

PC405648-1G PC405648-250MG
APOSPC405648-1GEA 261 EUR
APOSPC405648-1G APOSPC405648-250MG
3-(Pentafluorothio)thiophenol
3-(Pentafluorothio)thiophenol
Formula: C₆H₅F₅S₂
Storage Temperature: Refrigerator
MDL Number: MFCD27664885
CAS Number: 1820618-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR