(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine

Supplier: Apollo Scientific

PC408386-500MG
APOSPC408386-500MGEA 865 EUR
APOSPC408386-500MG
(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine
(4-Fluoro-benzyl)-[1-(4-fluoro-benzyl)-1H-benzoimidazol-2-yl]-amine
Formula: C₂₁H₁₇F₂N₃
MW: 349.377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09697750
CAS Number: 899809-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR