tert-Butyl 2-(((5-fluoropyrimidin-2-yl)oxy)methyl)pyrrolidine-1-carboxylate

Supplier: Apollo Scientific

PC408418-500MG
APOSPC408418-500MGEA 553 EUR
APOSPC408418-500MG
tert-Butyl 2-(((5-fluoropyrimidin-2-yl)oxy)methyl)pyrrolidine-1-carboxylate
tert-Butyl 2-(((5-fluoropyrimidin-2-yl)oxy)methyl)pyrrolidine-1-carboxylate
Formula: C₁₄H₂₀FN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD22577017
CAS Number: 1420870-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR