1-Nitro-2-(pentafluoroethyl)benzene

Supplier: Apollo Scientific

PC408830-1G
APOSPC408830-1GEA 1580 EUR
APOSPC408830-1G
1-Nitro-2-(pentafluoroethyl)benzene
1-Nitro-2-(pentafluoroethyl)benzene
Formula: C₈H₄F₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD28122884
CAS Number: 60979-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR