N-Boc-trans-4-fluoro-L-proline methyl ester

Supplier: Apollo Scientific

Synonyms: (2S,4R)-1-tert-Butyl 2-methyl 4-fluoropyrrolidine-1,2-dicarboxylate

PC409009-1G PC409009-250MG
APOSPC409009-1GEA 71.2 EUR
APOSPC409009-1G APOSPC409009-250MG
N-Boc-trans-4-fluoro-L-proline methyl ester
N-Boc-trans-4-fluoro-L-proline methyl ester
Formula: C₁₁H₁₈FNO₄
Storage Temperature: Ambient
MDL Number: MFCD07367993
CAS Number: 203866-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR