trans-4-Fluoro-L-Proline methyl ester hydrochloride 95+%

Supplier: Apollo Scientific

PC409019-250MG
APOSPC409019-250MGEA 132 EUR
APOSPC409019-250MG
trans-4-Fluoro-L-Proline methyl ester hydrochloride 95+%
trans-4-Fluoro-L-Proline methyl ester hydrochloride
Formula: C₆H₁₀FNO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD07374354
CAS Number: 58281-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR