(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

PC4098-1G PC4098-25G PC4098-5G
APOSPC4098-1GEA 21.9 EUR
APOSPC4098-1G APOSPC4098-25G APOSPC4098-5G
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid 98%
(3-Fluoro-4-(trifluoromethyl)phenyl)acetic acid
Formula: C₉H₆F₄O₂
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00236303
CAS Number: 238754-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR