3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile

Supplier: Apollo Scientific

PC410384-1G
APOSPC410384-1GEA 386 EUR
APOSPC410384-1G
3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
3-[1',5'-Dimethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
Formula: C₁₂H₁₂F₃N₅
Storage Temperature: Ambient
MDL Number: MFCD08559012
CAS Number: 1006333-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR