2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride

Supplier: Apollo Scientific
Warning

PC411329-1G PC411329-5G
APOSPC411329-1GEA 284 EUR
APOSPC411329-1G APOSPC411329-5G
2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride
2,2,2-Trifluoro-1-pyridin-2-ylethanamine dihydrochloride
Formula: C₇H₇F₃N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD16872039
CAS Number: 1241675-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR